Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Hydroxyphenylboronic acid pinacol ester
4-Hydroxyphenylboronic acid pinacol ester 98%
4-Hydroxyphenylboronic acid pinacol ester 98%
:
Apollo Scientific
4-Hydroxyphenylboronic acid pinacol ester 98%
:
Apollo Scientific
:
269409-70-3
:
:
C₁₂H₁₇BO₃
:
MFCD02093756
:
220.08 g/mol
:
113…117 °C
:
GHS07
:
Ambient