Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Hydroxyphenylboronic acid pinacol ester
2-Hydroxyphenylboronic acid pinacol ester 98%
2-Hydroxyphenylboronic acid pinacol ester 98%
:
Apollo Scientific
2-Hydroxyphenylboronic acid pinacol ester 98%
:
Apollo Scientific
:
269409-97-4
:
:
C₁₂H₁₇BO₃
:
MFCD02093754
:
220.08 g/mol
:
282 °C
:
1.05 g/cm³ (25 °C)
:
GHS07
:
Ambient