
Ethyl-5-amino-1-phenyl-1H-pyrazole-4-carboxylate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:16078-71-0
:
Formula:C₇H₁₁N₃O₂
MDL Number:MFCD00020731
:231.26 g/mol
Melting Pt:98…104 °C
:GHS07
Storage Temperature:Ambient