Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Chloro-m-toluic acid
4-Chloro-m-toluic acid
4-Chloro-m-toluic acid
Irritant
4-Chloro-m-toluic acid
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-Chloro-m-toluic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
7697-29-2
:
Formula:
C₈H₇ClO₂
MDL Number:
MFCD00045853
:
170.6 g/mol
Melting Pt:
212…216 °C
:
GHS07
Storage Temperature:
Ambient