Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Aminoimidazole hemisulphate
2-Aminoimidazole hemisulphate
2-Aminoimidazole hemisulphate
Irritant
2-Aminoimidazole hemisulphate
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-Aminoimidazole hemisulphate
Supplier:
Apollo Scientific
About this item
CAS Number:
1450-93-7
:
Formula:
C₆H₁₂N₆O₄S
MDL Number:
MFCD00013162
:
264.26 g/mol
Melting Pt:
270 °C (dec.)
EINECS:
215-918-9
:
GHS07
Storage Temperature:
Ambient