Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(Benzyloxy)phenylboronic acid
4-(Benzyloxy)phenylboronic acid 98%
4-(Benzyloxy)phenylboronic acid 98%
Irritant
4-(Benzyloxy)phenylboronic acid 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-(Benzyloxy)phenylboronic acid 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
146631-00-7
:
Formula:
C₁₃H₁₃BO₃
MDL Number:
MFCD01075705
:
228.06 g/mol
Melting Pt:
198…200 °C
:
GHS07
Storage Temperature:
Refrigerator