
3-(Aminomethyl)phenylboronic acid pinacol ester, hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:380430-65-9
:
Formula:C₁₃H₂₁BClNO₂
MDL Number:MFCD03410294
:269.58 g/mol
Melting Pt:169…171 °C
:GHS07
Storage Temperature:Refrigerator