Order Entry
Finland
Orders LinkContactUsLinkComponent
 

2,6-Dichlorophenol 98%

Corrosive
Irritant
Environment
New Product
Supplier:   Apollo Scientific
2,6-Dichlorophenol 98%
Supplier:  Apollo Scientific
New Product
Certificates
2,6-Dichlorophenol 98%

2 Options Available Below

Product Details & Documents
  • CAS Number:87-65-0
  • Formula:C₆H₄Cl₂O
  • MDL Number:MFCD00002176
  • Molecular Weight:163 g/mol
  • Boiling Pt:218…220 °C
  • Melting Pt:64…68 °C
  • Storage Temperature:Ambient
  • EINECS:201-761-3
  • UN:2020
  • ADR:6.1,III
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 165,00 €
    165,00 €
    1 * 250 g
       165,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 474,00 €
    474,00 €
    1 * 1 kg
       474,00 €

    Specifications:
    New Product
    New Product