Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Chloropicolinic acid
6-Chloropicolinic acid 98%
6-Chloropicolinic acid 98%
Irritant
6-Chloropicolinic acid 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
6-Chloropicolinic acid 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
4684-94-0
:
Formula:
C₆H₄ClNO₂
MDL Number:
MFCD00155390
:
157.56 g/mol
Melting Pt:
185…194 °C
:
GHS07
Storage Temperature:
Ambient