Order Entry
Finland
ContactUsLinkComponent
5-Bromo-2-((tert-butoxycarbonyl)amino)benzoic acid 97%
Irritant
5-Bromo-2-((tert-butoxycarbonyl)amino)benzoic acid 97%
Catalog # APOSOR019848-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-Bromo-2-((tert-butoxycarbonyl)amino)benzoic acid 97%
Catalog # APOSOR019848-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :316.1500 g/mol
Melting Pt:180…183 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:306937-20-2
Formula:C₁₂H₁₄BrNO₄
MDL Number:MFCD01619006