Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Amino-3-nitrobenzonitrile
4-Amino-3-nitrobenzonitrile
4-Amino-3-nitrobenzonitrile
Irritant
4-Amino-3-nitrobenzonitrile
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-Amino-3-nitrobenzonitrile
Supplier:
Apollo Scientific
About this item
CAS Number:
6393-40-4
:
Formula:
C₇H₅N₃O₂
MDL Number:
MFCD00013373
:
163.14 g/mol
Melting Pt:
159…162 °C
Storage Temperature:
Ambient
UN:
3439
ADR:
6.1,III
:
GHS07