Gallium indium alloy (75.5:24.5 w%), eutectic
Catalog # APOSIN1880-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
Melting Pt:15.7 °C
Density:6.25 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:1760
ADR:8,III
:GHS05
CAS Number:56451-02-6
MDL Number:MFCD00144387