Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Barium sulphide
Barium sulphide 75%, Technical Grade
Barium sulphide 75%, Technical Grade
Barium sulphide 75%, Technical Grade
Supplier:
Apollo Scientific
Certificates
Expand image
Barium sulphide 75%, Technical Grade
Supplier:
Apollo Scientific
Certificates
About this item
CAS Number:
21109-95-5
:
Formula:
BaS
MDL Number:
MFCD00014192
Melting Pt:
1200 °C
Density:
4.25 g/cm³ (25 °C)
:
169.4 g/mol
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
Documentation