
Vitamin K1 (phytomenadione, phylloquinone)
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:84-80-0
:81818-54-4
Formula:C₃₁H₄₆O₂
MDL Number:MFCD00214063
:450.6958 g/mol
Boiling Pt:∼140 °C/0.001mmHg
Melting Pt:∼ …20 °C
Density:0.984 g/cm³ (25 °C)
Flash Pt:>110 °C
:GHS07
Storage Temperature:Refrigerator