Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,4-Diaminobutane dihydrochloride
1,4-Diaminobutane dihydrochloride
1,4-Diaminobutane dihydrochloride
:
Apollo Scientific
1,4-Diaminobutane dihydrochloride
:
Apollo Scientific
:
333-93-7
:
:
C₄H₁₄Cl₂N₂
:
MFCD00012526
:
280 °C (dec.)
:
161.07 g/mol
:
206-375-9
:
GHS07
:
Refrigerator