Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Paromomycin sulphate
Paromomycin sulphate
Paromomycin sulphate
Paromomycin sulphate
Supplier:
Apollo Scientific
Certificates
Expand image
Paromomycin sulphate
Supplier:
Apollo Scientific
About this item
CAS Number:
1263-89-4
:
Formula:
C₂₃H₄₅N₅O₁₄·H₂SO₄
MDL Number:
MFCD00079278
:
615.6 g/mol
:
GHS07
Storage Temperature:
Ambient