Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Bisindolylmaleimide I
Bisindolylmaleimide I
Bisindolylmaleimide I
Irritant
Bisindolylmaleimide I
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Bisindolylmaleimide I
Supplier:
Apollo Scientific
About this item
CAS Number:
133052-90-1
:
Formula:
C₂₅H₂₄N₄O₂
MDL Number:
MFCD00236428
:
412.48 g/mol
:
GHS07
Storage Temperature:
Freezer