Guanidinium thiocyanate ≥99.0%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:593-84-0
:
Formula:CH₅N₃·CHNS
MDL Number:MFCD00013027
:118.16 g/mol
Melting Pt:117…122 °C
Density:1.29 g/cm³ (25 °C)
Storage Temperature:Refrigerator
EINECS:209-812-1
UN:1759
ADR:8,III
:GHS05
:GHS07