Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Methylumbelliferyl beta-D-ribofuranoside
4-Methylumbelliferyl beta-D-ribofuranoside
4-Methylumbelliferyl beta-D-ribofuranoside
4-Methylumbelliferyl beta-D-ribofuranoside
APOSBIM2050-50MG
:
Apollo Scientific
:
4-Methylumbelliferyl beta-D-ribofuranoside
APOSBIM2050-50MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
50 mg
Seq:
2370
:
:
308.29 g/mol
:
GHS07
:
Freezer
:
195385-93-4
:
C₁₅H₁₆O₇
:
MFCD06796941