
3-Maleimidopropionic acid
Catalog # APOSBIM1007-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Melting Pt:103…106 °C
:169.14 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:7423-55-4
Formula:C₇H₇NO₄
MDL Number:MFCD00043030