0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
EDTA tetrasodium salt dihydrate
EDTA tetrasodium salt dihydrate
EDTA tetrasodium salt dihydrate
:
Apollo Scientific
EDTA tetrasodium salt dihydrate
:
Apollo Scientific
:
10378-23-1
:
:
C₁₀H₁₂N₂Na₄O₈·2H₂O
:
MFCD00150026
:
>300 °C
:
416.2 g/mol
:
200-573-9
:
GHS05
:
GHS07
:
Ambient