Order Entry
Finland
ContactUsLinkComponent
Dextran sulphate sodium salt from leuconostoc 500,000
Dextran sulphate sodium salt from leuconostoc 500,000
Catalog # APOSBID1342-10G
Supplier:  Apollo Scientific
CAS Number:  
Dextran sulphate sodium salt from leuconostoc 500,000
Catalog # APOSBID1342-10G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :GHS07
Storage Temperature:Refrigerator
CAS Number:9011-18-1
Formula:(C₆H₇Na₃O₁₄S₃)n
MDL Number:MFCD00081551