Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Deoxy-L(+)-ribose
2-Deoxy-L-ribose
2-Deoxy-L-ribose
Irritant
2-Deoxy-L-ribose
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-Deoxy-L-ribose
Supplier:
Apollo Scientific
About this item
CAS Number:
18546-37-7
:
Formula:
C₅H₁₀O₄
MDL Number:
MFCD00132941
:
134.13 g/mol
Melting Pt:
69…72 °C
:
GHS07
Storage Temperature:
Freezer