5-CFDA (5-Carboxyfluorescein diacetate)
Catalog # APOSBIC1071-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:460.39 g/mol
Storage Temperature:Freezer
CAS Number:79955-27-4
Formula:C₂₅H₁₆O₉
MDL Number:MFCD00036872