Order Entry
Finland
ContactUsLinkComponent
Benzyl N-(2-aminoethyl)carbamate hydrochloride ≥99%
Benzyl N-(2-aminoethyl)carbamate hydrochloride ≥99%
Catalog # APOSBIBP1042-5G
Supplier:  Apollo Scientific
CAS Number:  
Benzyl N-(2-aminoethyl)carbamate hydrochloride ≥99%
Catalog # APOSBIBP1042-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :230.69 g/mol
Melting Pt:169…172 °C
Storage Temperature:Ambient
CAS Number:18807-71-1
 :72080-83-2
Formula:C₁₀H₁₄N₂O₂
MDL Number:MFCD00270150