(±)-1,2-Octanediol
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 1,2-Octanediol, (±)-1,2-Octylene glycol, (±)-1,2-Dihydroxy octane
- CAS Number:1117-86-8
- Formula:C₈H₁₈O₂
- MDL Number:MFCD00010738
- Molecular Weight:146.23 g/mol
- Boiling Pt:131…132 °C
- Melting Pt:28…31 °C
- Flash Pt:138 °C
- Storage Temperature:Ambient
3 Options Available Below
Product Details & Documents
Synonyms: 1,2-Octanediol, (±)-1,2-Octylene glycol, (±)-1,2-Dihydroxy octane
- CAS Number:1117-86-8
- Formula:C₈H₁₈O₂
- MDL Number:MFCD00010738
- Molecular Weight:146.23 g/mol
- Boiling Pt:131…132 °C
- Melting Pt:28…31 °C
- Flash Pt:138 °C
- Storage Temperature:Ambient



