Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Methyl-DL-tryptophan
4-Methyl-DL-tryptophan
4-Methyl-DL-tryptophan
4-Methyl-DL-tryptophan
Supplier:
Apollo Scientific
Certificates
Expand image
4-Methyl-DL-tryptophan
Supplier:
Apollo Scientific
About this item
CAS Number:
1954-45-6
:
Formula:
C₁₂H₁₄N₂O₂
MDL Number:
MFCD00055987
:
218.256 g/mol
EINECS:
217-785-2
:
GHS07
Storage Temperature:
Refrigerator