Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Bialaphos sodium
Bialaphos sodium
Bialaphos sodium
Bialaphos sodium
Supplier:
Apollo Scientific
Certificates
Expand image
Bialaphos sodium
Supplier:
Apollo Scientific
About this item
CAS Number:
71048-99-2
:
Formula:
C₁₁H₂₁N₃NaO₆P
MDL Number:
MFCD01674432
:
345.26 g/mol
:
GHS07
Storage Temperature:
Freezer