Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Azauracil
6-Azauracil
6-Azauracil
Irritant
6-Azauracil
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
6-Azauracil
Supplier:
Apollo Scientific
About this item
CAS Number:
461-89-2
:
Formula:
C₃H₃N₃O₂
MDL Number:
MFCD00006456
:
113.08 g/mol
Melting Pt:
274…275 °C
EINECS:
207-318-0
:
GHS07
Storage Temperature:
Ambient