Order Entry
Finland
Orders LinkContactUsLinkComponent
Agarose, low melting

Agarose, low melting

Irritant
Supplier:   Apollo Scientific
Agarose, low melting
Supplier:  Apollo Scientific
Certificates
Agarose, low melting

2 Options Available Below

Product Details & Documents

Low melting agaroses are synthetically derived, their low melting temperature allows for the recovery of undamaged nucleic acids below the denaturation temperature. The low gelling temperature ensures that the agarose will be in a liquid state at a temperature where in-gel manipulations can be performed without prior extraction of DNA from the gel slice.

  • Gel strength at 1.5% >500g/cm2
  • Gelling temp. 24-28°C
  • Melting temp. <65.5°C
  • Moisture <7%
  • Ash <0.4%
  • EEO <0.12
  • Sulphate <0.12%
  • Clarity (NTU) <4
  • DNAse/RNAse activity not detected.
  • CAS Number:9012-36-6
  • MDL Number:MFCD00081294
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 111,00 €
    111,00 €
    1 * 50 g
       111,00 €

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 192,00 €
    192,00 €
    1 * 100 g
       192,00 €