0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
Agarose
Agarose low EEO type D1, Molecular Biology Grade
Agarose low EEO type D1, Molecular Biology Grade
:
Apollo Scientific
Agarose low EEO type D1, Molecular Biology Grade
:
Apollo Scientific
:
9012-36-6
:
:
MFCD00081294
:
232-731-8
:
GHS07
:
Ambient