Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(S)-(+)-2-Amino-1-propanol
(S)-(+)-2-Amino-1-propanol 99%
(S)-(+)-2-Amino-1-propanol 99%
:
Apollo Scientific
(S)-(+)-2-Amino-1-propanol 99%
:
Apollo Scientific
:
2749-11-3
:
:
C₃H₉NO
:
MFCD00064412
:
75.11 g/mol
:
174…176 °C
:
0.965 g/cm³ (25 °C)
:
63 °C
:
Ambient
:
220-388-7
:
2735
:
8,II
:
GHS05