Order Entry
Finland
ContactUsLinkComponent
1,3-Benzenedimethanethiol 98%
1,3-Benzenedimethanethiol 98%
Catalog # APOSBI7567-5G
Supplier:  Apollo Scientific
CAS Number:  
1,3-Benzenedimethanethiol 98%
Catalog # APOSBI7567-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :170.29 g/mol
Storage Temperature:Ambient
CAS Number:41563-69-3
Formula:C₈H₁₀S₂
MDL Number:MFCD00012311