Order Entry
Finland
ContactUsLinkComponent
2-Acetamido-4-fluorobenzeneboronic acid, pinacol ester 95%
2-Acetamido-4-fluorobenzeneboronic acid, pinacol ester 95%
Catalog # APOSPC6998-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Acetamido-4-fluorobenzeneboronic acid, pinacol ester 95%
Catalog # APOSPC6998-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :279.115 g/mol
Storage Temperature:Ambient
CAS Number:1150271-67-2
Formula:C₁₄H₁₉BFNO₃
MDL Number:MFCD12026077