Order Entry
Finland
Orders LinkContactUsLinkComponent
 

5-Fluoro-2-methoxy-4-methylaniline

Irritant
Supplier:   Apollo Scientific
5-Fluoro-2-methoxy-4-methylaniline
Supplier:  Apollo Scientific
Certificates
5-Fluoro-2-methoxy-4-methylaniline

2 Options Available Below

Product Details & Documents
  • CAS Number:1692806-14-6
  • Formula:C₈H₁₀FNO
  • MDL Number:MFCD30338944
  • Molecular Weight:155.17 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 125,00 €
    125,00 €
    1 * 250 mg
       125,00 €

  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 368,00 €
    368,00 €
    1 * 1 g
       368,00 €