Order Entry
Finland
ContactUsLinkComponent
 
Novec HFE-71IPA Engineered Fluid (3M)
Catalog # APOSPC5527-100G
Supplier:  Apollo Scientific
CAS Number:  
Novec HFE-71IPA Engineered Fluid (3M)
Catalog # APOSPC5527-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
Boiling Pt:55 °C
Melting Pt:–42 °C
Density:1.42 g/cm³ (25 °C)
Storage Temperature:Ambient
CAS Number:185045-85-6
MDL Number:MFCD23704319