Methyl 5-(difluoromethyl)-2,4-dimethyl-1H-pyrrole-3-carboxylate
Catalog # APOSPC520172-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:203.186 g/mol
Storage Temperature:Ambient
Formula:C₉H₁₁F₂NO₂
MDL Number:MFCD29767652