4-(Difluoromethoxy)-2-nitroaniline 97%
Catalog # APOSPC501715-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:204.131 g/mol
Storage Temperature:Ambient
CAS Number:97963-76-3
Formula:C₇H₆F₂N₂O₃
MDL Number:MFCD09037370