Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Fluoro-2-nitrobenzotrifluoride
3-Fluoro-2-nitrobenzotrifluoride 95%
3-Fluoro-2-nitrobenzotrifluoride 95%
Toxic
Irritant
3-Fluoro-2-nitrobenzotrifluoride 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Toxic
Irritant
3-Fluoro-2-nitrobenzotrifluoride 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
1214335-98-4
:
Formula:
C₇H₃F₄NO₂
MDL Number:
MFCD12032180
:
209.098 g/mol
UN:
2810
ADR:
6.1,III
:
GHS06
:
GHS07
Storage Temperature:
Ambient