Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 2-(bromomethyl)-4-fluorobenzoate
Methyl 2-(bromomethyl)-4-fluorobenzoate
Methyl 2-(bromomethyl)-4-fluorobenzoate
Methyl 2-(bromomethyl)-4-fluorobenzoate
Supplier:
Apollo Scientific
Certificates
Expand image
Methyl 2-(bromomethyl)-4-fluorobenzoate
Supplier:
Apollo Scientific
About this item
CAS Number:
157652-28-3
:
Formula:
C₉H₈BrFO₂
MDL Number:
MFCD11505951
:
247.061 g/mol
Storage Temperature:
Refrigerator
UN:
3265
ADR:
8,III
:
GHS05