Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-3-fluorobenzyl bromide
2-Chloro-3-fluorobenzyl bromide
2-Chloro-3-fluorobenzyl bromide
APOSPC499012-1G
:
Apollo Scientific
:
2-Chloro-3-fluorobenzyl bromide
APOSPC499012-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
223.47 g/mol
:
41…44 °C
:
91 °C
:
Ambient
:
3261
:
8,III
:
GHS05
:
874285-19-5
:
C₇H₅BrClF
:
MFCD09038462