
2-Bromo-6-fluorobenzyl bromide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1548-81-8
:
Formula:C₇H₅Br₂F
MDL Number:MFCD10698675
:267.921 g/mol
Boiling Pt:128…131 °C (25 mm Hg)
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05