Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Fluoro-3-nitrobenzyl chloride
2-Fluoro-3-nitrobenzyl chloride 97%
2-Fluoro-3-nitrobenzyl chloride 97%
2-Fluoro-3-nitrobenzyl chloride 97%
Supplier:
Apollo Scientific
Certificates
Expand image
2-Fluoro-3-nitrobenzyl chloride 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
1020718-00-6
:
Formula:
C₇H₅ClFNO₂
MDL Number:
MFCD09955448
:
189.572 g/mol
Storage Temperature:
Ambient