2-(3-Bromo-4-fluorophenyl)-4-(trifluoromethyl)-1H-imidazole
Catalog # APOSPC450027-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:309.058 g/mol
Storage Temperature:Ambient
CAS Number:1309602-29-6
Formula:C₁₀H₅BrF₄N₂
MDL Number:MFCD19442113