Order Entry
Finland
ContactUsLinkComponent
(R)-2,2,2-Trifluoro-N-(1-phenylethyl)acetamide 95%
(R)-2,2,2-Trifluoro-N-(1-phenylethyl)acetamide 95%
Catalog # APOSPC421193-1G
Supplier:  Apollo Scientific
CAS Number:  
(R)-2,2,2-Trifluoro-N-(1-phenylethyl)acetamide 95%
Catalog # APOSPC421193-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :217.188 g/mol
Storage Temperature:Ambient
CAS Number:39995-50-1
Formula:C₁₀H₁₀F₃NO
MDL Number:MFCD00192525