trans-3-Amino-1-(trifluoromethyl)cyclobutan-1-ol hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1408075-16-0
:
Formula:C₅H₈F₃NO·ClH
MDL Number:MFCD23106128
:191.579 g/mol
Storage Temperature:Ambient
Frequently Bought Together