1-(1,5-Dimethyl-1H-pyrazol-4-yl)-N-(4-fluorobenzyl)methanamine
Catalog # APOSPC410467-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:233.285 g/mol
Storage Temperature:Ambient
CAS Number:1006465-11-7
Formula:C₁₃H₁₆FN₃
MDL Number:MFCD04968207