N-[(1,3-Dimethyl-1H-pyrazol-4-yl)methyl]-3-(trifluoromethyl)aniline
Catalog # APOSPC410394-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:269.266 g/mol
Storage Temperature:Ambient
CAS Number:1006336-83-9
Formula:C₁₃H₁₄F₃N₃
MDL Number:MFCD04968305