Order Entry
Finland
ContactUsLinkComponent
1-[(3-Fluorophenoxy)methyl]-3,5-dimethyl-1H-pyrazol-4-amine
1-[(3-Fluorophenoxy)methyl]-3,5-dimethyl-1H-pyrazol-4-amine
Catalog # APOSPC410310-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[(3-Fluorophenoxy)methyl]-3,5-dimethyl-1H-pyrazol-4-amine
Catalog # APOSPC410310-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :235.257 g/mol
Storage Temperature:Ambient
CAS Number:1004192-82-8
Formula:C₁₂H₁₄FN₃O
MDL Number:MFCD04967714