[1-(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-pyrrolidin-3-yl]-carbamic acid tert-butyl ester
Catalog # APOSPC408387-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:365.778 g/mol
Storage Temperature:Ambient
CAS Number:1354448-68-2
Formula:C₁₅H₁₉ClF₃N₃O₂
MDL Number:MFCD20921630